Barium chloride dihydrate ≥99.997% (metals basis), Puratronic®

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Barium dichloride

10646.14 10646.22 10646.36
ALFA10646.14EA 0 SGD
ALFA10646.14 ALFA10646.22 ALFA10646.36
Barium chloride dihydrate ≥99.997% (metals basis), Puratronic®
Barium chloride dihydrate

Purity: 99.997% CAS No.: 10326-27-9 Molecular Formula: BaCl2.2H2O Formula Weight: 244.28 (208.25anhy) Physical Form: Powder Density: 3.097 Appearance: White Odour: Odourless Melting Point: 113° -2H2O Merck Reference: 14,971 UN No.: 1564 MDL No.: MFCD00149154

Formula: BaCl₂·2H₂O
Boiling Pt: 1560 °C (1013 hPa)
Melting Pt: 113 °C (-2H₂O)
Density: 3,097
Storage Temperature: Ambient
MDL Number: MFCD00149154
CAS Number: 10326-27-9
EINECS: 600-412-6
UN: 1564
ADR: 6.1,III
Merck Index: 14,971

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR