Chromium(III) chloride hexahydrate ≥99.998% (metals basis), Puratronic®

Supplier: Thermo Fisher Scientific
Danger

10686.06 10686.14
ALFA10686.06EA 0 SGD
ALFA10686.06 ALFA10686.14
Chromium(III) chloride hexahydrate ≥99.998% (metals basis), Puratronic®
Chromium (III) chloride hexahydrate

Purity: 99.998% CAS No.: 10060-12-5 Molecular Formula: Cl3Cr.6H2O Formula Weight: 266.45 (158.35anhy) Physical Form: Crystalline Aggregates Density: 1.835 Appearance: Dark green Odour: Odourless Melting Point: 83° Merck Reference: 14,2222 UN No.: 3260 MDL No.: MFCD00149660

Formula: CrCl₃·6H₂O
Melting Pt: 83 °C
Density: 1,835
Storage Temperature: Ambient
MDL Number: MFCD00149660
CAS Number: 10060-12-5
EINECS: 233-038-3
UN: 3260
ADR: 8,III
Merck Index: 14,2222

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR