Lead(II) acetate trihydrate 99,995 Metals basis, Puratronic®

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Acetic acid lead (II) salt trihydrate, Lead diacetate trihydrate

10719.06 10719.18 10719.30
ALFA10719.06EA 36.9 SGD
ALFA10719.06 ALFA10719.18 ALFA10719.30
Lead(II) acetate trihydrate 99,995 Metals basis, Puratronic®
Lead (II) acetate trihydrate

Purity: 99.995% CAS No.: 6080-56-4 Molecular Formula: Pb(OOCCH3)2.3H2O Formula Weight: 379.33 (325.28anhy) Physical Form: Crystalline Density: 2.55 Melting Point: 75° Beilstein Reference: 3730298 Merck Reference: 14,5397 UN No.: 1616 MDL No.: MFCD00150023

Formula: (H₃CCOO)₂Pb·3H₂O
MW: 379.34 g/mol
Boiling Pt: 280
Melting Pt: 75
Density: 2.55
Storage Temperature: Ambient
MDL Number: MFCD00150023
CAS Number: 6080-56-4
EINECS: 206-104-4
UN: 1616
ADR: 6.1 III
Merck Index: 14,5397
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=351511a44fbb598a2d7fa1fa4ab395427b6168f1&vid=67e858e5f0dc4d04dd796541b3c0ad5aa73f8dc8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR