Potassium sulphate 99,997 Metals basis, Puratronic®

Supplier: Thermo Fisher Scientific

10845.06 10845.14
ALFA10845.06EA 0 SGD
ALFA10845.06 ALFA10845.14
Potassium sulphate 99,997 Metals basis, Puratronic®
Potassium sulphate

Purity: 99.997% CAS No.: 7778-80-5 Molecular Formula: K2O4S Formula Weight: 174.27 Physical Form: Powder Density: 2.662 Appearance: White Odour: Odourless Melting Point: 1069° Boiling Point: 1689° Merck Reference: 14,7674 MDL No.: MFCD00011388

Formula: K₂SO₄
MW: 174.26 g/mol
Boiling Pt: 1689 °C
Melting Pt: 1069
Density: 2.662
Storage Temperature: Ambient
MDL Number: MFCD00011388
CAS Number: 7778-80-5
EINECS: 231-915-5
Merck Index: 14,7674

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR