Rhodium(III) chloride hydrate (38,5 - 45,5% Rh)

Supplier: Thermo Fisher Scientific
Danger

11032.03 11032.06 11032.14 11032.01
ALFA11032.03EA 0 SGD
ALFA11032.03 ALFA11032.06 ALFA11032.14 ALFA11032.01
Rhodium(III) chloride hydrate (38,5 - 45,5% Rh)
Rhodium (III) chloride hydrate

CAS No.: 20765-98-4 Molecular Formula: RhCl3.xH2O Formula Weight: 209.26(anhy) Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: Dark Red Melting Point: 100° dec. Solubility: Soluble in alcohol, water. Slightly soluble in acetone Merck Reference: 14,8188 MDL No.: MFCD00149839

Formula: Cl₃Rh·H₂O
Melting Pt: 100 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00149839
CAS Number: 20765-98-4
EINECS: 233-165-4
Merck Index: 14,8188

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR