Diboron trioxide, dehydrated (max. 0.02% H₂O) ≥99.999% (metals basis), Puratronic®

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Boron oxide, Boric anhydride

11159.QE 11159.QH
ALFA11159.QEEA 0 SGD
ALFA11159.QE ALFA11159.QH
Diboron trioxide, dehydrated (max. 0.02% H₂O) ≥99.999% (metals basis), Puratronic®
di-Boron trioxide

Purity: 99.999% CAS No.: 1303-86-2 Molecular Formula: B2O3 Formula Weight: 69.62 Physical Form: Cylinder, 3.7cm dia, 32g/cylinder Density: 2.46 +/- 0.01 Appearance: White Odour: Odourless Melting Point: ca 450° Boiling Point: 1860° Merck Reference: 14,1337 MDL No.: MFCD00011315


In the electronics industry as a liquid encapsulent for Czochralski-type growth of III-V semiconductor single crystals (e.g., GaAs, GaP, InP)

Formula: B₂O₃
MW: 69.62 g/mol
Boiling Pt: 1860 °C
Melting Pt: ∼450 °C
Density: 2.46
Storage Temperature: Ambient
MDL Number: MFCD00011315
CAS Number: 1303-86-2
EINECS: 215-125-8
REACH: 01-2119486655-24
Merck Index: 14,1337

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR