Ytterbium(III) chloride hydrate ≥99.99% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Fisher Scientific
Warning

11194.09 11194.18
ALFA11194.09EA 0 SGD
ALFA11194.09 ALFA11194.18
Ytterbium(III) chloride hydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Ytterbium (III) chloride hydrate

Purity: 99.99% CAS No.: 19423-87-1 Molecular Formula: YbCl3.xH2O (x~6) Formula Weight: 279.40(anhy) Physical Form: Crystalline Aggregates Density: 2.58 Appearance: Green Odour: Odourless Melting Point: 150-155° Merck Reference: 14,10105 MDL No.: MFCD00149885

Formula: Cl₃Yb·H₂O
Melting Pt: 150…155 °C
Density: 2,58
Storage Temperature: Ambient
MDL Number: MFCD00149885
CAS Number: 19423-87-1
EINECS: 233-800-5
Merck Index: 14,10105

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR