Lanthanum(III) chloride heptahydrate ≥99.99% (REO, rare earth oxide basis), REacton®

Supplier: Thermo Fisher Scientific
Warning

11268.18 11268.30 11268.A1
ALFA11268.18EA 0 SGD
ALFA11268.18 ALFA11268.30 ALFA11268.A1
Lanthanum(III) chloride heptahydrate ≥99.99% (REO, rare earth oxide basis), REacton®
Lanthanum (III) chloride heptahydrate

Purity: 99.99% CAS No.: 10025-84-0 Molecular Formula: LaCl3.7H2O Formula Weight: 371.37 (245.27anhy) Physical Form: Wet Crystalline Aggregates Odour: Odourless Merck Reference: 14,5363 MDL No.: MFCD00149756

Formula: LaCl₃·7H₂O
Boiling Pt: ∼860 °C (1013 hPa)
Melting Pt: 91 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00149756
CAS Number: 10025-84-0
EINECS: 233-237-5
Merck Index: 14,5363

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR