Sodium carbonate, anhydrous ≥99.5% ACS

Supplier: Thermo Fisher Scientific
Warning

11552.18 11552.36 11552.A3
ALFA11552.18EA 0 SGD
ALFA11552.18 ALFA11552.36 ALFA11552.A3
Sodium carbonate, anhydrous ≥99.5% ACS
Sodium carbonate

Purity: 99.5% min CAS No.: 497-19-8 Molecular Formula: Na2CO3 Formula Weight: 105.99 Physical Form: Granular Density: 2.53 Refractive Index: 1.535 Sensitivity: Hygroscopic Odour: Odourless pH: 11,5 Solubility: Soluble in water. Insoluble in alcohol Beilstein Reference: 4154566 Merck Reference: 14,8596 MDL No.: MFCD00003494

Formula: Na₂CO₃
Boiling Pt: 1600 °C (1013 hPa)
Melting Pt: 854 °C
Density: 2,53
Storage Temperature: Ambient
MDL Number: MFCD00003494
CAS Number: 497-19-8
EINECS: 207-838-8
REACH: 01-2119485498-19
Merck Index: 14,8596
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6ff30f1487cdef86add98b38bfa8eb3dac14f6&vid=c5b129b7acb5c897b9d970ea3130d61fb1934b54&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR