Rhodium(III) chloride, anhydrous 99,9 Metals basis Rh ≥ 48.7%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Rhodium(III) chloride, Rhodium trichloride

11815.01 11815.03 11815.06
ALFA11815.01EA 0 SGD
ALFA11815.01 ALFA11815.03 ALFA11815.06
Rhodium(III) chloride, anhydrous 99,9 Metals basis Rh ≥ 48.7%
Rhodium (III) chloride

Purity: 99.9% CAS No.: 10049-07-7 Molecular Formula: Cl3Rh Formula Weight: 209.26 Physical Form: Powder Sensitivity: Hygroscopic Appearance: Red-brown Odour: Odourless Merck Reference: 14,8188 MDL No.: MFCD00011204

Formula: Cl₃Rh
MW: 209.26 g/mol
Density: 5.38 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011204
CAS Number: 10049-07-7
EINECS: 233-165-4
Merck Index: 14,8188

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR