Ammonium heptamolybdate tetrahydrate 81-83% (MoO₃ basis) ACS

Supplier: Thermo Fisher Scientific
Warning

11831.22 11831.18 11831.36 11831.A3
ALFA11831.22EA 0 SGD
ALFA11831.22 ALFA11831.18 ALFA11831.36 ALFA11831.A3
Ammonium heptamolybdate tetrahydrate 81-83% (MoO₃ basis) ACS
Ammonium heptamolybdate tetrahydrate

Purity: 81-83% CAS No.: 12054-85-2 Molecular Formula: H24Mo7N6O24.4H2O Formula Weight: 1235.86 (1163.79anhy) Physical Form: Powder Density: 2.498 Appearance: White to pale green Melting Point: ca 190° dec. Solubility: Soluble in water (1:2 ratio). Insoluble in alcohol Merck Reference: 14,533 MDL No.: MFCD00167059


In photography, analytical lab reagent for phosphate, lead, arsenate determination

Formula: (NH₄)₆Mo₇O₂₄·4H₂O
Melting Pt: ∼ 190 °C (dec.)
Density: 2.498
Storage Temperature: Ambient
MDL Number: MFCD00167059
CAS Number: 12054-85-2
EINECS: 234-722-4
Merck Index: 14,533

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR