Indium(III) chloride, anhydrous ≥99.999% (metals basis)

Supplier: Thermo Fisher Scientific
Danger

11856.03 11856.09 11856.18 11856.30
ALFA11856.03EA 0 SGD
ALFA11856.03 ALFA11856.09 ALFA11856.18 ALFA11856.30
Indium(III) chloride, anhydrous ≥99.999% (metals basis)
Indium (III) chloride

Purity: 99.999% CAS No.: 10025-82-8 Molecular Formula: InCl3 Formula Weight: 221.18 Physical Form: Crystalline Density: 3.460 Appearance: White Odour: Odourless Melting Point: 586° Boiling Point: 800° Merck Reference: 14,4958 UN No.: 3260 MDL No.: MFCD00011058

Formula: Cl₃In
Boiling Pt: 800 °C
Melting Pt: 586 °C
Density: 3,460
Storage Temperature: Ambient
MDL Number: MFCD00011058
CAS Number: 10025-82-8
EINECS: 233-043-0
UN: 3260
ADR: 8,II
Merck Index: 14,4958

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR