Diboron trioxide ≥97.5%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Boron oxide, Boric anhydride

12290.36 12290.A3 12290.A9
ALFA12290.36EA 0 SGD
ALFA12290.36 ALFA12290.A3 ALFA12290.A9
Diboron trioxide ≥97.5%
di-Boron trioxide

Purity: 97.5% min CAS No.: 1303-86-2 Molecular Formula: B2O3 Formula Weight: 69.62 Physical Form: -60 Mesh Density: 2.46 +/- 0.01 Appearance: White Odour: Odourless Melting Point: ca 450° Boiling Point: 1860° Solubility: Slightly soluble in water @ 25°C. Soluble in water @ 100°C Merck Reference: 14,1337 MDL No.: MFCD00011315


As a flux, acid catalyst, or chemical intermediate in production of boron halides, esters, carbide, nitride, and metallic borides

Formula: B₂O₃
MW: 69.62 g/mol
Boiling Pt: 1860 °C
Melting Pt: ∼450 °C
Density: 2.46
Storage Temperature: Ambient
MDL Number: MFCD00011315
CAS Number: 1303-86-2
EINECS: 215-125-8
REACH: 01-2119486655-24
Merck Index: 13,01327

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR