Barium chloride dihydrate ACS

Supplier: Thermo Fisher Scientific
Danger

12310.36 12310.A3
ALFA12310.36EA 0 SGD
ALFA12310.36 ALFA12310.A3
Barium chloride dihydrate ACS
Barium chloride dihydrate

CAS No.: 10326-27-9 Molecular Formula: BaCl2.2H2O Formula Weight: 244.28 (208.25anhy) Physical Form: Crystalline Density: 3.097 Odour: Odourless Merck Reference: 14,971 UN No.: 1564 MDL No.: MFCD00149154

Formula: BaCl₂·2H₂O
Boiling Pt: 1560 °C (1013 hPa)
Melting Pt: 113 °C (-2H₂O)
Density: 3,097
Storage Temperature: Ambient
MDL Number: MFCD00149154
CAS Number: 10326-27-9
EINECS: 233-788-1
UN: 1564
ADR: 6.1,III
Merck Index: 14,971

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR