Chromium(III) chloride, anhydrous 98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Chromium trichloride

12336.18 12336.30
ALFA12336.18EA 0 SGD
ALFA12336.18 ALFA12336.30
Chromium(III) chloride, anhydrous 98%
Chromium (III) chloride

Purity: 98% CAS No.: 10025-73-7 Molecular Formula: CrCl3 Formula Weight: 158.35 Physical Form: Crystalline Density: 2.87 Appearance: Violet Boiling Point: 1300(subl) Merck Reference: 14,2222 UN No.: 3260 MDL No.: MFCD00010948

Formula: Cl₃Cr
MW: 158.35 g/mol
Boiling Pt: 1300 °C (subl.)
Melting Pt: 1152
Density: 2.87
Storage Temperature: Ambient
MDL Number: MFCD00010948
CAS Number: 10025-73-7
EINECS: 233-038-3
UN: 3260
ADR: 8 III
Merck Index: 14,2222

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR