Iron(III) chloride, anhydrous 98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Iron trichloride, Ferric chloride

12357.09 12357.22 12357.A1 12357.A7 12357.C4
ALFA12357.09EA 0 SGD
ALFA12357.09 ALFA12357.22 ALFA12357.A1 ALFA12357.A7 ALFA12357.C4
Iron(III) chloride, anhydrous 98%
Iron (III) chloride

Purity: 98% CAS No.: 7705-08-0 Molecular Formula: Cl3Fe Formula Weight: 162.21 Physical Form: Crystalline Density: 2.90 Appearance: Black Odour: Odourless pH: 1.8 Melting Point: 306° Boiling Point: 316° Solubility: Freely soluble in water, alcohol, ether, acetone. Slightly soluble in CS2. Insoluble in ethyl acetate Merck Reference: 14,4019 UN No.: 1773 MDL No.: MFCD00011005

Formula: FeCl₃
MW: 162.21 g/mol
Boiling Pt: 316 °C
Melting Pt: 306 °C
Density: 2.90
Storage Temperature: Ambient
MDL Number: MFCD00011005
CAS Number: 7705-08-0
EINECS: 231-729-4
UN: 1773
ADR: 8 III
Merck Index: 14,4019

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR