Aluminium oxide ≥99% (metals basis)

Supplier: Thermo Fisher Scientific

Synonyms: Aluminium(III) oxide, Alumina C, Aluminum oxide

12553.A3
ALFA12553.A3EA 0 SGD
ALFA12553.A3
Aluminium oxide ≥99% (metals basis)
Aluminium oxide

Purity: 99% CAS No.: 1344-28-1 Molecular Formula: Al2O3 Formula Weight: 101.96 Physical Form: Fine Powder Density: 3.965 Refractive Index: 1.768 Appearance: White Odour: Odourless Melting Point: 2045° Boiling Point: 2980° Merck Reference: 14,356 MDL No.: MFCD00003424


Catalyst support, chemical sensors, as absorbent for gases and water vapors, manufacturing of abrasives, refractories, ceramics, electrical insulators and resistors, paper, crucibles, as chromatographic matrix, in fluxes, light bulbs, heat-resistant fibers

Formula: Al₂O₃
Boiling Pt: 2980 °C
Melting Pt: 2045 °C
Density: 3,965
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
REACH: 01-2119529248-35
Merck Index: 14,356

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR