Bis(dibenzylideneacetone)palladium

Supplier: Thermo Fisher Scientific

Synonyms: Bis(dibenzylideneacetone)palladium(0)

Argon_filled

12764.01 12764.03 12764.06
ALFA12764.01EA 0 SGD
ALFA12764.01 ALFA12764.03 ALFA12764.06
Bis(dibenzylideneacetone)palladium
Bis(dibenzylideneacetone)palladium

CAS No.: 32005-36-0 Molecular Formula: C34H28O2Pd Formula Weight: 575.00 Physical Form: Powder, Packaged under argon Sensitivity: Air and Moisture Sensitive Appearance: Brown to purple Melting Point: 135° dec. Solubility: Slightly soluble in CH2Cl2, CHCl3 and benzene MDL No.: MFCD00051942


Applications: Hydrogenation, isomerization, carbonylation, oxidation, C-C bond formation

Formula: C₃₄H₂₈O₂Pd
MW: 575 g/mol
Melting Pt: 135 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00051942
CAS Number: 32005-36-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR