Calcium sulphide ≥99.9% (metals basis excluding Sr) (max. 0.05% Sr)

Supplier: Thermo Fisher Scientific
Warning

13121.06 13121.14
ALFA13121.06EA 0 SGD
ALFA13121.06 ALFA13121.14
Calcium sulphide ≥99.9% (metals basis excluding Sr) (max. 0.05% Sr)
Calcium sulphide

Purity: 99.9% CAS No.: 20548-54-3 Molecular Formula: CaS Formula Weight: 72.14 Physical Form: -325 Mesh Powder Density: 2.5 Melting Point: dec. Merck Reference: 14,1707 UN No.: 3134 MDL No.: MFCD00015990

Formula: CaS
Melting Pt: Dec.
Density: 2,5
Storage Temperature: Ambient
MDL Number: MFCD00015990
CAS Number: 20548-54-3
EINECS: 243-873-5
UN: 3134
ADR: 4.3,III
Merck Index: 14,1707

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR