Barium chloride, ultra dry ≥99.998% (metals basis)

Supplier: Thermo Fisher Scientific
Danger


ampouled under argon

13686.14
ALFA13686.14EA 0 SGD
ALFA13686.14
Barium chloride, ultra dry ≥99.998% (metals basis)
Barium chloride

Purity: 99.998% CAS No.: 10361-37-2 Molecular Formula: BaCl2 Formula Weight: 208.25 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 3.9 Appearance: White Odour: Odourless Melting Point: 963° Boiling Point: 1560° Merck Reference: 14,971 UN No.: 1564 MDL No.: MFCD00003445

Formula: BaCl₂
Boiling Pt: 1560 °C
Melting Pt: 963 °C
Density: 3,9
Storage Temperature: Ambient
MDL Number: MFCD00003445
CAS Number: 10361-37-2
EINECS: 233-788-1
UN: 1564
ADR: 6.1,III
Merck Index: 14,971

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR