Chloropentaamminecobalt(III) chloride

Supplier: Thermo Fisher Scientific
Warning

13765.04 13765.09 13765.18
ALFA13765.04EA 0 SGD
ALFA13765.04 ALFA13765.09 ALFA13765.18
Chloropentaamminecobalt(III) chloride
Chloropentaamminecobalt (III) chloride

Slightly soluble in water, soluble in conc. H₂SO₄. Insoluble in alcohol


CAS No.: 13859-51-3 Molecular Formula: H15Cl3CoN5 Formula Weight: 250.44 Physical Form: Powder Density: 1.819 Appearance: Purple Odour: Odourless Melting Point: dec. Solubility: Slightly soluble in water, soluble in conc. H2SO4. Insoluble in alcohol MDL No.: MFCD00064728

Formula: H₁₅Cl₃CoN₅
Melting Pt: Dec.
Density: 1,819
Storage Temperature: Ambient
MDL Number: MFCD00064728
CAS Number: 13859-51-3
EINECS: 237-594-8

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR