Sodium periodate ≥98%

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Sodium metaperiodate

13798.09 13798.22 13798.36 13798.A3
ALFA13798.09EA 0 SGD
ALFA13798.09 ALFA13798.22 ALFA13798.36 ALFA13798.A3
Sodium periodate ≥98%
Sodium periodate

Purity: 98% CAS No.: 7790-28-5 Molecular Formula: INaO4 Formula Weight: 213.89 Physical Form: Powder Density: 3.86 Appearance: White Odour: Odourless Melting Point: 300° dec. Solubility: Soluble in water and acids Merck Reference: 14,8640 UN No.: 1479 MDL No.: MFCD00003534


Source of periodic acid, as an analytical reagent, and as an oxidizing agent

Formula: NaIO₄
MW: 213.89 g/mol
Boiling Pt: 300
Melting Pt: 300 °C (dec.)
Density: 3.86
Storage Temperature: Ambient
MDL Number: MFCD00003534
CAS Number: 7790-28-5
EINECS: 232-197-6
UN: 1479
ADR: 5.1 II
Merck Index: 13,08714

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR