Vanadium(III) chloride 99 Metals basis

Supplier: Thermo Fisher Scientific
Danger

14039.09 14039.18
ALFA14039.09EA 0 SGD
ALFA14039.09 ALFA14039.18
Vanadium(III) chloride 99 Metals basis
Vanadium (III) chloride

Purity: 99% CAS No.: 7718-98-1 Molecular Formula: VCl3 Formula Weight: 157.30 Physical Form: -4 Mesh Density: 3.00 Appearance: Pink to purple Odour: Acrid UN No.: 2475 MDL No.: MFCD00011454

Formula: Cl₃V
MW: 157.3 g/mol
Density: 3.00
Storage Temperature: Ambient
MDL Number: MFCD00011454
CAS Number: 7718-98-1
EINECS: 231-744-6
UN: 2475
ADR: 8 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR