Potassium sulphate 99,0 ACS

Supplier: Thermo Fisher Scientific

14311.36 14311.A3
ALFA14311.36EA 0 SGD
ALFA14311.36 ALFA14311.A3
Potassium sulphate 99,0 ACS
Potassium sulphate

Purity: 99.0% min CAS No.: 7778-80-5 Molecular Formula: K2SO4 Formula Weight: 174.27 Physical Form: Crystalline Density: 2.662 Melting Point: 1069° Boiling Point: 1689° Solubility: Soluble in water. Insoluble in alcohol Merck Reference: 14,7674 MDL No.: MFCD00011388

Formula: K₂SO₄
MW: 174.26 g/mol
Boiling Pt: 1689 °C
Melting Pt: 1069
Density: 2.662
Storage Temperature: Ambient
MDL Number: MFCD00011388
CAS Number: 7778-80-5
EINECS: 231-915-5
Merck Index: 14,7674

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR