Barium carbonate ≥99.8%

Supplier: Thermo Fisher Scientific
Warning

14341.30
ALFA14341.30EA 0 SGD
ALFA14341.30
Barium carbonate ≥99.8%
Barium carbonate

Purity: 99.8% CAS No.: 513-77-9 Molecular Formula: BaCO3 Formula Weight: 197.35 Physical Form: 1 Micron Powder Density: 4.43 Melting Point: 811° Boiling Point: dec. Merck Reference: 14,969 MDL No.: MFCD00003448

Formula: BaCO₃
Boiling Pt: Dec.
Melting Pt: 811 °C
Density: 4,43
Storage Temperature: Ambient
MDL Number: MFCD00003448
CAS Number: 513-77-9
EINECS: 208-167-3
UN: 1564
ADR: 6.1,III
Merck Index: 14,969

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR