Lithium chloride, ultra dry ≥99.9% (metals basis)

Supplier: Thermo Fisher Scientific
Warning


ampouled under argon

14540.09 14540.18 14540.30
ALFA14540.09EA 0 SGD
ALFA14540.09 ALFA14540.18 ALFA14540.30
Lithium chloride, ultra dry ≥99.9% (metals basis)
Lithium chloride

Purity: 99.9% CAS No.: 7447-41-8 Molecular Formula: LiCl Formula Weight: 42.39 Physical Form: -10 Mesh Beads, Ampouled under argon Density: 2.068 Melting Point: 605° Boiling Point: 1325-1360° Merck Reference: 14,5528 MDL No.: MFCD00011078

Formula: LiCl
Boiling Pt: 1325…1360 °C
Melting Pt: 605 °C
Density: 2,068
Storage Temperature: Ambient
MDL Number: MFCD00011078
CAS Number: 7447-41-8
EINECS: 231-212-3
Merck Index: 14,5528

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR