Azomethine H hydrate

Supplier: Thermo Fisher Scientific

16810.04 16810.09 16810.14
ALFA16810.04EA 125 SGD
ALFA16810.04 ALFA16810.09 ALFA16810.14
Azomethine H hydrate
Azomethine H hydrate

CAS No.: 32266-60-7 Molecular Formula: C17H13NO8S2.xH2O Formula Weight: 423.40(anhy) Physical Form: Crystalline MDL No.: MFCD00066536

Formula: C₁₇H₁₃NO₈S₂·H₂O
Storage Temperature: Ambient
MDL Number: MFCD00066536
CAS Number: 32266-60-7
EINECS: 250-975-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR