Bismuth(III) chloride, anhydrous 99,9 Metals basis

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Bismuth(III) chloride, Bismuth trichloride

17115.14 17115.22
ALFA17115.14EA 0 SGD
ALFA17115.14 ALFA17115.22
Bismuth(III) chloride, anhydrous 99,9 Metals basis
Bismuth (III) chloride

Purity: 99.9% CAS No.: 7787-60-2 Molecular Formula: BiCl3 Formula Weight: 315.34 Physical Form: Powder Density: 4.75 Appearance: White Melting Point: 230-232° Boiling Point: ca 430° subl. Solubility: Soluble in HCl, HNO3, acetone, ethyl acetate. Insoluble in alcohol. Decomposed by water to BiOCl Merck Reference: 14,1262 UN No.: 3260 MDL No.: MFCD00003461


Catalysts for organic reactions, preparation of Bi salts

Formula: BiCl₃
MW: 315.34 g/mol
Boiling Pt: ∼430 °C (subl.)
Melting Pt: 230…232 °C
Density: 4.75
Storage Temperature: Ambient
MDL Number: MFCD00003461
CAS Number: 7787-60-2
EINECS: 232-123-2
UN: 3260
ADR: 8 III
Merck Index: 14,1262

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR