Bis(2-ethylhexyl)phosphate 95

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Bis(2-ethylhexyl) hydrogen phosphate, Di-(2-ethylhexyl)phosphoric acid

17723.22 17723.36 17723.A3
ALFA17723.22EA 0 SGD
ALFA17723.22 ALFA17723.36 ALFA17723.A3
Bis(2-ethylhexyl)phosphate 95
Bis(2-ethylhexyl) phosphate

Purity: 95% CAS No.: 298-07-7 Molecular Formula: C16H35O4P Formula Weight: 322.43 Flash Point: 171°(340°F) Physical Form: Liquid Density: 0.965 Refractive Index: 1.4420 Appearance: Colourless pH: 2-3 Melting Point: -50° Boiling Point: 48°/12mm UN No.: 1902 MDL No.: MFCD00009492


Extraction of metals

Formula: C₁₆H₃₅O₄P
MW: 322.43 g/mol
Boiling Pt: 48 °C (12 mmHg)
Melting Pt: –50 °C
Density: 0.965
Flash Pt: 171 °C (340 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009492
CAS Number: 298-07-7
EINECS: 206-056-4
UN: 1902
ADR: 8 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR