Yttrium(III) chloride, anhydrous 99,9 REO basis

Supplier: Thermo Fisher Scientific
Warning

18682.18 18682.09 18682.30
ALFA18682.18EA 0 SGD
ALFA18682.18 ALFA18682.09 ALFA18682.30
Yttrium(III) chloride, anhydrous 99,9 REO basis
Yttrium (III) chloride

Purity: 99.9% CAS No.: 10361-92-9 Molecular Formula: Cl3Y Formula Weight: 195.26 Physical Form: -20 Mesh Density: 2.67 Appearance: White to pale yellow Odour: Odourless Melting Point: 721° Merck Reference: 14,10107 MDL No.: MFCD00011470

Formula: Cl₃Y
MW: 195.26 g/mol
Boiling Pt: 1488
Melting Pt: 721 °C
Density: 2.67
Storage Temperature: Ambient
MDL Number: MFCD00011470
CAS Number: 10361-92-9
EINECS: 233-801-0
Merck Index: 14,10107

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR