Potassium sulphate 99,99 Metals basis

Supplier: Thermo Fisher Scientific

30485.06 30485.14 30485.22
ALFA30485.06EA 0 SGD
ALFA30485.06 ALFA30485.14 ALFA30485.22
Potassium sulphate 99,99 Metals basis
Potassium sulphate

Purity: 99.99% CAS No.: 7778-80-5 Molecular Formula: K2SO4 Formula Weight: 174.27 Physical Form: Powder Density: 2.662 Melting Point: 1069° Boiling Point: 1689° Merck Reference: 14,7674 MDL No.: MFCD00011388

Formula: K₂SO₄
MW: 174.26 g/mol
Boiling Pt: 1689 °C
Melting Pt: 1069
Density: 2.662
Storage Temperature: Ambient
MDL Number: MFCD00011388
CAS Number: 7778-80-5
EINECS: 231-915-5
Merck Index: 14,7674

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR