n-Undecane 99

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Undecane

31807.18 31807.30
ALFA31807.18EA 0 SGD
ALFA31807.18 ALFA31807.30
n-Undecane 99
n-Undecane

Purity: 99% CAS No.: 1120-21-4 Molecular Formula: C11H24 Formula Weight: 156.30 Flash Point: 60°(140°F) Physical Form: Liquid Density: 0.74 Refractive Index: 1.4173 Appearance: Colourless Odour: Mild Melting Point: -26° Boiling Point: 196° UN No.: 2330 MDL No.: MFCD00008959

Formula: H₃C(CH₂)₉CH₃
MW: 156.31 g/mol
Boiling Pt: 196 °C
Melting Pt: –26 °C
Density: 0.74
Flash Pt: 60 °C (140 °F)
Storage Temperature: Ambient
MDL Number: MFCD00008959
CAS Number: 1120-21-4
EINECS: 214-300-6
UN: 2330
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR