Gallium trifluoride, anhydrous ≥99.85% (metals basis)

Supplier: Thermo Fisher Scientific
Danger

32112.06
ALFA32112.06EA 0 SGD
ALFA32112.06
Gallium trifluoride, anhydrous ≥99.85% (metals basis)
Gallium trifluoride

Purity: 99.85% CAS No.: 7783-51-9 Molecular Formula: GaF3 Formula Weight: 126.72 Physical Form: Powder Density: 4.47 Appearance: White Odour: Odourless Melting Point: 800° subl. Boiling Point: ca 1000° Merck Reference: 14,4354 UN No.: 3260 MDL No.: MFCD00011021

Formula: F₃Ga
MW: 126.72 g/mol
Boiling Pt: ∼1000 °C
Melting Pt: 800 °C (subl.)
Density: 4.47
Storage Temperature: Ambient
MDL Number: MFCD00011021
CAS Number: 7783-51-9
EINECS: 232-004-5
UN: 3260
ADR: 8 III
Merck Index: 14,4354

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR