Insulating beads, ceramic, AL23
Supplier: Thermo Fisher Scientific
32915.KX
ALFA32915.KXEA 0 SGD
ALFA32915.KX
Insulating beads, ceramic, AL23
Beads Ceramic Beads
Pure Al₂O₃ Aluminium oxide, AL23.
Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.