Ceramic plates, rectangular or square, AL23
Supplier: Thermo Fisher Scientific
33155.KF 33156.KF 33158.KF
ALFA33155.KFEA 0 SGD
ALFA33155.KF ALFA33156.KF ALFA33158.KF
Ceramic plates, rectangular or square, AL23
Trays/Dishes/Pots Laboratory Trays/Dishes
Pure Al₂O₃ Aluminium oxide AL23.
Dense, excellent thermal and electrical resistance properties, corrosion resistant, permeable to microwaves.