Aluminium oxide cement ≥95% (SiO₂ ∼ 5%)

Supplier: Thermo Fisher Scientific

Synonyms: Aluminium(III) oxide, Alumina C, Aluminum oxide

33201.A1
ALFA33201.A1EA 0 SGD
ALFA33201.A1
Aluminium oxide cement ≥95% (SiO₂ ∼ 5%)
Aluminium oxide

CAS No.: 1344-28-1 Molecular Formula: Al2O3 Physical Form: Agglomerates 150 micron Odour: Odourless MDL No.: MFCD00211795

Formula: Al₂O₃
Boiling Pt: 2980 °C (1013 hPa)
Melting Pt: 2045 °C
Density: 3,94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
REACH: 01-2119529248-35
Merck Index: 13,00355

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR