Sodium carbonate solution_standardised 0.05 N aqueous solution

Supplier: Thermo Fisher Scientific

Synonyms: Calcined soda, Carbonic acid disodium salt

35609.K2
ALFA35609.K2EA 0 SGD
ALFA35609.K2
Sodium carbonate solution_standardised 0.05 N aqueous solution
Sodium carbonate

CAS No.: 497-19-8 Molecular Formula: Na2CO3 Formula Weight: 105.99 Physical Form: Liquid MDL No.: MFCD00003494

Formula: Na₂CO₃
MW: 105.99 g/mol
Boiling Pt: 1600
Melting Pt: 854
Storage Temperature: Ambient
MDL Number: MFCD00003494
CAS Number: 497-19-8
EINECS: 207-838-8
Merck Index: 14,08596
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a6ff30f1487cdef86add98b38bfa8eb3dac14f6&vid=c5b129b7acb5c897b9d970ea3130d61fb1934b54&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR