Oxalic acid 0.05 mol/l (0.1 N) in aqueous solution volumetric solution

Supplier: Thermo Fisher Scientific

Synonyms: Ethanedioic acid

35622.K2
ALFA35622.K2EA 0 SGD
ALFA35622.K2
Oxalic acid 0.05 mol/l (0.1 N) in aqueous solution volumetric solution
Oxalic acid

Molecular Formula: HO2CCO2H Formula Weight: 90.04 Physical Form: Liquid Appearance: Colourless Odour: Odourless MDL No.: MFCD00002573

Formula: C₂H₂O₄
Storage Temperature: Ambient
MDL Number: MFCD00002573
CAS Number: 144-62-7
EINECS: 205-634-3
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR