Cerium(IV) sulphate standardised solution 0.1 N in aqueous solution

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Cerium disulphate, Ceric sulphate, Cerium(IV) sulphate

35650.K2
ALFA35650.K2EA 0 SGD
ALFA35650.K2
Cerium(IV) sulphate standardised solution 0.1 N in aqueous solution
Cerium (IV) sulphate
Formula: Ce(SO₄)₂
MW: 332.24 g/mol
Melting Pt: 195
Density: 1.05
Storage Temperature: Ambient
MDL Number: MFCD00148852
CAS Number: 13590-82-4
EINECS: 237-029-5
UN: 3264
ADR: 8 III
Merck Index: 14,1990

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR