Chromium(III) chloride, anhydrous 99,9 Metals basis

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Chromium trichloride

35691.06 35691.14 35691.22
ALFA35691.06EA 0 SGD
ALFA35691.06 ALFA35691.14 ALFA35691.22
Chromium(III) chloride, anhydrous 99,9 Metals basis
Chromium (III) chloride

Purity: 99.9% CAS No.: 10025-73-7 Molecular Formula: CrCl3 Formula Weight: 158.35 Physical Form: Flake Density: 2.87 Appearance: Violet Melting Point: ca 1150° Merck Reference: 14,2222 UN No.: 3260 MDL No.: MFCD00010948

Formula: Cl₃Cr
MW: 158.35 g/mol
Melting Pt: ∼ 1150 °C
Density: 2.87
Storage Temperature: Ambient
MDL Number: MFCD00010948
CAS Number: 10025-73-7
EINECS: 233-038-3
UN: 3260
ADR: 8 III
Merck Index: 14,2222

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR