Europium(III) chloride, ultra dry ≥99.99% (REO, rare earth oxide basis)

Supplier: Thermo Fisher Scientific
Warning


ampouled under argon

35738.03 35738.06 35738.14
ALFA35738.03EA 0 SGD
ALFA35738.03 ALFA35738.06 ALFA35738.14
Europium(III) chloride, ultra dry ≥99.99% (REO, rare earth oxide basis)
Europium (III) chloride

Purity: 99.99% CAS No.: 10025-76-0 Molecular Formula: EuCl3 Formula Weight: 258.32 Physical Form: Powder, Ampouled under argon Sensitivity: Hygroscopic Solubility: Soluble in water Merck Reference: 14,3904 MDL No.: MFCD00010994

Formula: Cl₃Eu
Melting Pt: 850 °C
Density: 4,89 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010994
CAS Number: 10025-76-0
EINECS: 233-040-4
Merck Index: 14,3904

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR