Lithium chloride ≥99% ACS

Supplier: Thermo Fisher Scientific
Warning

36217.36 36217.A3
ALFA36217.36EA 0 SGD
ALFA36217.36 ALFA36217.A3
Lithium chloride ≥99% ACS
Lithium chloride

Purity: 99% min CAS No.: 7447-41-8 Molecular Formula: LiCl Formula Weight: 42.39 Physical Form: Granular Density: 2.068 Melting Point: 605° Boiling Point: 1325-1360° Solubility: Very soluble in water, alcohol, ether, pyridine, and nitrobenzene Merck Reference: 14,5528 MDL No.: MFCD00011078


In air-conditioning, welding and soldering flux, dry batteries, heat-exchange media, salt baths, and desiccants

Formula: LiCl
Boiling Pt: 1325…1360 °C
Melting Pt: 605 °C
Density: 2,068
Storage Temperature: Ambient
MDL Number: MFCD00011078
CAS Number: 7447-41-8
EINECS: 231-212-3
Merck Index: 14,5528

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR