Cobalt(II) chloride hexahydrate 98.0-102.0% ACS

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Cobalt Chloride, Hexahydrate, Cobalt dichloride hexahydrate, Cobalt Chloride, 6-Hydrate, Cobaltous chloride hexahydrate

36554.22 36554.36
ALFA36554.22EA 0 SGD
ALFA36554.22 ALFA36554.36
Cobalt(II) chloride hexahydrate 98.0-102.0% ACS
Cobalt (II) chloride hexahydrate

Purity: 98.0-102.0% CAS No.: 7791-13-1 Molecular Formula: Cl2Co.6H2O Formula Weight: 237.93 (129.84anhy) Physical Form: Crystalline Density: 1.924 Appearance: Red-purple Odour: Odourless pH: 4.9 Melting Point: 87° Solubility: Soluble in water, alcohol, acetone, ether, glycerol Merck Reference: 14,2437 UN No.: 3077 MDL No.: MFCD00149652

Formula: CoCl₂·6H₂O
Boiling Pt: 110 °C (1013 hPa)
Melting Pt: 87 °C
Density: 1,924
Storage Temperature: Ambient
MDL Number: MFCD00149652
CAS Number: 7791-13-1
EINECS: 231-589-4
UN: 3077
ADR: 9,III
Merck Index: 14,2437

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR