Yttrium(III) 2-ethylhexanoate ≥99.8% (metals basis)

Supplier: Thermo Fisher Scientific
Warning

36661.14
ALFA36661.14EA 0 SGD
ALFA36661.14
Yttrium(III) 2-ethylhexanoate ≥99.8% (metals basis)
Yttrium (III) 2-ethylhexanoate

Purity: 99.8% CAS No.: 103470-68-4 Molecular Formula: C24H45O6Y Formula Weight: 518.16 Physical Form: Powder

Formula: C₂₄H₄₅O₆Y
Storage Temperature: Ambient
MDL Number: MFCD00070441
CAS Number: 103470-68-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR