Chromium(III) potassium sulphate dodecahydrate 98.0-102.0% ACS

Supplier: Thermo Fisher Scientific
Warning

36715.22 36715.36
ALFA36715.22EA 0 SGD
ALFA36715.22 ALFA36715.36
Chromium(III) potassium sulphate dodecahydrate 98.0-102.0% ACS
Chromium (III) potassium sulphate dodecahydrate

Purity: 98.0-102.0% CAS No.: 7788-99-0 Molecular Formula: CrKO8S2.12H2O Formula Weight: 499.39 (283.21anhy) Physical Form: Solid Density: 1.83 Appearance: Dark purple Odour: Odourless Melting Point: 89° Merck Reference: 14,2229 MDL No.: MFCD00149917

Formula: CrK(SO₄)₂·12H₂O
Melting Pt: 89 °C
Density: 1,83
Storage Temperature: Ambient
MDL Number: MFCD00149917
CAS Number: 7788-99-0
EINECS: 233-401-6
Merck Index: 14,2229

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR