Methyl cellulose, 4000 mPa.S (2% H₂O), powder

Supplier: Thermo Fisher Scientific

Synonyms: Cellulose methyl ether

36718.22 36718.36 36718.A4
ALFA36718.22EA 0 SGD
ALFA36718.22 ALFA36718.36 ALFA36718.A4
Methyl cellulose, 4000 mPa.S (2% H₂O), powder
Methyl cellulose

CAS No.: 9004-67-5 Physical Form: Powder Odour: Odourless Merck Reference: 14,6040 MDL No.: MFCD00081763


Caution: Incompatible with strong oxidising agents, bleach, perchloric acid, nitric acid, perchlorates, alkali nitrates, alkali nitrites and calcium oxide.

Storage Temperature: Ambient
MDL Number: MFCD00081763
CAS Number: 9004-67-5
Merck Index: 14,6040

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR