Titanium(III) chloride aluminum(III) chloride TiCl3_76.0-78.5

Supplier: Thermo Fisher Scientific
Danger

Synonyms: Titanium(III) chloride, Al reduced, Titanium aluminum chloride, Titanium(III) chloride, Aluminum titanium chloride

36729.22 36729.36
ALFA36729.22EA 0 SGD
ALFA36729.22 ALFA36729.36
Titanium(III) chloride aluminum(III) chloride TiCl3_76.0-78.5
Titanium(III) chloride aluminum(III) chloride

CAS No.: 12003-13-3 Molecular Formula: (TiCl3)3AlCl3 Formula Weight: 596.15 Physical Form: Powder Sensitivity: Air and Moisture Sensitive UN No.: UN3260 MDL No.: MFCD00145412

Formula: (TiCl₃)₃AlCl₃
MW: 596.15 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00145412
CAS Number: 12003-13-3
EINECS: 234-421-8
ADR: 8 II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR