Bis[2-carboxyethylgermanium(IV)] sesquioxide 99,7

Supplier: Thermo Fisher Scientific
Warning

36758.06 36758.14 36758.22
ALFA36758.06EA 0 SGD
ALFA36758.06 ALFA36758.14 ALFA36758.22
Bis[2-carboxyethylgermanium(IV)] sesquioxide 99,7
Bis[2-carboxyethylgermanium(IV)] sesquioxide

Purity: 99.7% CAS No.: 12758-40-6 Molecular Formula: C6H10Ge2O7 Formula Weight: 339.25 Physical Form: Powder Odour: Odourless Melting Point: 320° dec. MDL No.: MFCD00049319

Formula: C₆H₁₀Ge₂O₇
MW: 339.40 g/mol
Melting Pt: 320 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00049319
CAS Number: 12758-40-6
EINECS: 235-800-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR