Silicon standard solution, 1000 mg/L Si in water with hydrofluoric acid (max. 1%) (from (NH₄)₂SiF₆), standard for ICP, Specpure®

Supplier: Thermo Fisher Scientific

38717.AD 38717.AE 38717.AP
ALFA38717.ADEA 0 SGD
ALFA38717.AD ALFA38717.AE ALFA38717.AP
Silicon standard solution, 1000 mg/L Si in water with hydrofluoric acid (max. 1%) (from (NH₄)₂SiF₆), standard for ICP, Specpure®
Standards Cationic Standards Silicium Standards

Molecular Formula: (NH₄)₂SiF₆ in H₂O/tr. F-
Physical Form: Liquid
MDL No.: MFCD00085311


Certifications: Traceable to NIST

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR