Crucibles, glassy carbon
Supplier: Thermo Fisher Scientific
39011.KT 38376.KT 39003.KT 39004.KT 39005.KT 39006.KT 39007.KT 39008.KT 39010.KT 39012.KT 39014.KT
ALFA39011.KTEA 0 SGD
ALFA39011.KT ALFA38376.KT ALFA39003.KT ALFA39004.KT ALFA39005.KT ALFA39006.KT ALFA39007.KT ALFA39008.KT ALFA39010.KT ALFA39012.KT ALFA39014.KT
Crucibles, glassy carbon
Crucibles
These crucibles are manufactured from glassy carbon. Available in conical and cylindrical types.
  • Resistance to all wet decomposition agents
  • No contamination of analytical samples
  • Stability to acid and alkaline melts
  • No wetting effect of metal melts
  • Good resistance to thermal shock allows rapid heating and cooling times
  • Resistive to abrasive wear
  • Good electrical conductivity
Glassy carbon, a brittle form of carbon with a randomised structure, has certain specific properties making it appropriate for fields of application outside the scope of carbonMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable